Learn More
MSC Baldwin Filters Fuel Element, 20/45 Micron, 2 1/4 in OD
Catalog No. 5000509660
Encompass
This fuel element is designed to keep fuel clean, ensuring engines run at maximum efficiency by filtering out contaminants. It is suitable for a variety of applications, including automotive, agriculture, and industrial settings.
  • Provides 20 micron nominal and 45 micron absolute filtration.
  • Features an attached gasket for secure installation.
  • Has an outside diameter of 2 1/4 inches (57.2 mm).
  • Has an inside diameter of 1 1/16 inches (27.0 mm) on one end.
  • Measures 3 31/32 inches (100.8 mm) in length.
  • Designed for use in Bennett fuel pumps.
Catalog No. 50-005-09660 Supplier MSC Supplier No. 55868863
This fuel element is designed to keep fuel clean, ensuring engines run at maximum efficiency by filtering out contaminants. It is suitable for a variety of applications, including automotive, agriculture, and industrial settings.
  • Provides 20 micron nominal and 45 micron absolute filtration.
  • Features an attached gasket for secure installation.
  • Has an outside diameter of 2 1/4 inches (57.2 mm).
  • Has an inside diameter of 1 1/16 inches (27.0 mm) on one end.
  • Measures 3 31/32 inches (100.8 mm) in length.
  • Designed for use in Bennett fuel pumps.