Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PF9 tetrasodium salt, Tocris Bioscience™

Catalog No. 40741
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-741 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 40-741 Supplier R&D Systems Supplier No. 4074/1
This item is not returnable. View return policy

Potent GPR17 agonist

Potent agonist of GPR17 (EC50 = 36 pM).

Chemical Identifiers

CAS 851265-78-6
Molecular Formula C18H16N5Na4O13P3
Molecular Weight (g/mol) 695.228
InChI Key ONTPGXYSNYMMOK-YEJXUIBNSA-J
Synonym pf9 tetrasodium salt, 2-phenylethynyl adenosine-5'-triphosphate tetrasodium salt
PubChem CID 90488909
IUPAC Name tetrasodium;[[[(2R,3S,4R)-5-[6-amino-2-(2-phenylethynyl)purin-9-yl]-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl]oxy-oxidophosphoryl] phosphate
SMILES C1=CC=C(C=C1)C#CC2=NC3=C(C(=N2)N)N=CN3C4C(C(C(O4)COP(=O)([O-])OP(=O)([O-])OP(=O)([O-])[O-])O)O.[Na+].[Na+].[Na+].[Na+]

Specifications

Quantity 1 mg
Formula Weight 695.23
Percent Purity >98%
Chemical Name or Material PF9 tetrasodium salt
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.