Learn More
MSC Baldwin Fuel Filter Element, Primary, with Bail Handle, 9.8 Micron Nominal
Catalog No. 5000508417
Encompass
Modern fuel injection systems require fuel to be free of both particulate and water contamination. This fuel filter element ensures fuel is clean, helping engines run at maximum efficiency.
  • Primary fuel element with bail handle
  • Effectively removes particulate and water contamination from fuel
  • Optimizes engine performance and efficiency
  • Features a 9.8 nominal; 27 absolute micron rating
  • Compatible with various equipment including Allis Chalmers, Ingersoll-Rand, International, and Kohler Generators
Catalog No. 50-005-08417 Supplier MSC Supplier No. 55865992
Modern fuel injection systems require fuel to be free of both particulate and water contamination. This fuel filter element ensures fuel is clean, helping engines run at maximum efficiency.
  • Primary fuel element with bail handle
  • Effectively removes particulate and water contamination from fuel
  • Optimizes engine performance and efficiency
  • Features a 9.8 nominal; 27 absolute micron rating
  • Compatible with various equipment including Allis Chalmers, Ingersoll-Rand, International, and Kohler Generators