Learn More
Siemens Healthineers PFA-100 Instrument (US/CA)
Catalog No. 23044716 Shop All Siemens Healthineers Products
Change view
Click to view available options
Sample Type:
Citrated Whole Blood
Catalog No. Sample Type
23-044-716 Citrated Whole Blood
1 options
This item is not returnable. View return policy
Catalog No. 23-044-716 Supplier Siemens Healthineers Supplier No. 10459298
This product is not orderable online. For ordering, please contact your Fisher Healthcare Sales Representative.
This item is not returnable. View return policy
The PFA-100™ System measures the complex process of primary hemostasis and aids in the rapid detection of platelet dysfunction. It is the first commercially available in vitro testing system to incorporate high-shear flow in which the process of platelet adhesion and aggregation following a vascular injury is simulated in-vitro.
The PFA-100™ System provides automated assessment for inherited, acquired, or drug-induced platelet dysfunction.
  • Helps detect inherited, acquired and drug-induced platelet dysfunction
  • Screening test for patients with impaired primary hemostasis, such as von Willebrand disease
  • Verifies efficacy of desmopressin acetate (DDAVP) therapy in pre-surgical patients
  • Assesses the platelet dysfunction due to the effect of Aspirin
  • Evaluates platelet dysfunction in children over 1 year of age
  • Evaluates platelet dysfunction in multiple clinical settings, such as presurgical screening, high-risk pregnancy and menorrhagia
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis System
Sample Type Citrated Whole Blood
Throughput Single Test: Col/EPI or Col/ADP: 6 tests/hour; Duplicate Test: Col/EPI or Col/ADP: 10 tests/hour
Operating System High-shear force dynamic flow system
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov