Learn More
Perkin Elmer US LLC PFA 47 mm M-200 Spray Chamber for Avio 200/500
Catalog No. 502105390
Encompass_Preferred
Constructed entirely from chemically inert PFA, the PureChamber is available in low flow format for maximum aerosol transport and high flow format, ideal for ICP-OES applications. The PureCap accepts any 6 mm nebulizer but is ideally suited for the PFA MicroFlow nebulizer providing an O-Ring free PFA sample introduction system. The PFA 47 mm M-200 Spray Chamber with Gas Port (N0791848) is available separately.
Catalog No. 50-210-5390 Supplier Perkin Elmer US LLC Supplier No. N0791847
Add to Cart
Add to Cart
Request a Quote
Constructed entirely from chemically inert PFA, the PureChamber is available in low flow format for maximum aerosol transport and high flow format, ideal for ICP-OES applications. The PureCap accepts any 6 mm nebulizer but is ideally suited for the PFA MicroFlow nebulizer providing an O-Ring free PFA sample introduction system. The PFA 47 mm M-200 Spray Chamber with Gas Port (N0791848) is available separately.