Learn More
Perkin Elmer US LLC PFA Adapter for Glass Nebulizers
Catalog No. 502105208
Encompass_Preferred
The mixing lit kit for online addition of internal standard is an easy way to use internal standards without having to manually spike each sample with internal standard. The kit can also be used to mix any two reagents for other purposes. The low dead-volume, fluoropolymer mixing tee ensures good mixing at typical flow rates for a wide variety of liquids, including HF. Includes: Low dead-volume, fluoropolymer mixing tee 0.25 mm ID (green) for flow rates < 1 mL/min 0.5 mm ID (orange) for flow rates > 0.3 mL/min Internal standard probe and addition line Fluoropolymer peripump fittings Peristaltic pump tubing starter pack
Catalog No. 50-210-5208 Supplier Perkin Elmer US LLC Supplier No. N8145365
The mixing lit kit for online addition of internal standard is an easy way to use internal standards without having to manually spike each sample with internal standard. The kit can also be used to mix any two reagents for other purposes. The low dead-volume, fluoropolymer mixing tee ensures good mixing at typical flow rates for a wide variety of liquids, including HF. Includes: Low dead-volume, fluoropolymer mixing tee 0.25 mm ID (green) for flow rates < 1 mL/min 0.5 mm ID (orange) for flow rates > 0.3 mL/min Internal standard probe and addition line Fluoropolymer peripump fittings Peristaltic pump tubing starter pack