Learn More
Perkin Elmer US LLC PFA Barrel Spray Chamber with End Cap for NexION 300/350
Catalog No. 502104771
Encompass_Preferred
This barrel spray chamber, made of perfluoroalkoxy alkanes (PFA), is hydrofluoric acid-resistant. It is designed to provide high sensitivity for semiconductor and other high purity applications.
Catalog No. 50-210-4771 Supplier Perkin Elmer US LLC Supplier No. N8142000
Add to Cart
Add to Cart
Request a Quote
This barrel spray chamber, made of perfluoroalkoxy alkanes (PFA), is hydrofluoric acid-resistant. It is designed to provide high sensitivity for semiconductor and other high purity applications.