Learn More
Chemglass Life Sciences PFA Coated Stainless Steel Thermocouple: 1/2" Diameter x 45" Long, Type "T", Fits Reactor Capacity: 75L
SDP
Supplier:  Chemglass Life Sciences CG3498116
Encompass_Preferred
PFA Coated Stainless Steel Thermocouple: 1/4" Diameter x 26" Long, Type "T", Fits Reactor Capacity: 10L-20L. Thermocouples for use with Chemglass process reactors. The detachable thermocouples are for direct insertion in the reactors using a CG-1048-A or CG-1042-E thermocouple adapter. Detachable type thermocouples have a standard male plug (OST) and require a coiled extension cord for connection to the CG-3498 temperature monitor. The 4 long Hastelloy C-276 thermocouples are supplied with a 4ft FEP insulated cord having a standard male plug (OST).
Catalog No. 50-153-1624