Learn More
Chemglass Life Sciences PFA Coated Stainless Steel Thermocouple: 1/4" Diameter x 32" Long, Type "T", Fits Reactor Capacity: 30L & 50L Short
SDP
Catalog No. CG3498108
Encompass_Preferred
PFA Coated Stainless Steel Thermocouple: 1/4" Diameter x 32" Long, Type "T", Fits Reactor Capacity: 30L & 50L Short. Thermocouples for use with Chemglass process reactors. Available in both ¿T¿ (blue connections) and ¿K¿ (yellow connections). The detachable thermocouples are for direct insertion in the reactors using a CG-1048-A or CG-1042-E thermocouple adapter, while the 4¿ long Hastelloy C-276 style is for use with the CG-1971 baffles and adapters. Detachable type thermocouples have a standard male plug (OST) and require a coiled extension cord for connection to the CG-3498 temperature monitor. The 4¿ long Hastelloy C-276 thermocouples are supplied with a 4ft FEP insulated cord having a standard male plug (OST).PLEASE NOTE: THE 1/2¿ DIAMETER PROBES HAVE A TRANSITION TO 6¿ OF PTFE LEADS AND ARE CLEAR FEP PTFE COATED. Maximum Probe Temperature Based on Coating- FEP is 200¿C and the Black PFA is 250¿C
Catalog No. CG3498108 Supplier Chemglass Life Sciences Supplier No. CG3498108
PFA Coated Stainless Steel Thermocouple: 1/4" Diameter x 32" Long, Type "T", Fits Reactor Capacity: 30L & 50L Short. Thermocouples for use with Chemglass process reactors. Available in both ¿T¿ (blue connections) and ¿K¿ (yellow connections). The detachable thermocouples are for direct insertion in the reactors using a CG-1048-A or CG-1042-E thermocouple adapter, while the 4¿ long Hastelloy C-276 style is for use with the CG-1971 baffles and adapters. Detachable type thermocouples have a standard male plug (OST) and require a coiled extension cord for connection to the CG-3498 temperature monitor. The 4¿ long Hastelloy C-276 thermocouples are supplied with a 4ft FEP insulated cord having a standard male plug (OST).PLEASE NOTE: THE 1/2¿ DIAMETER PROBES HAVE A TRANSITION TO 6¿ OF PTFE LEADS AND ARE CLEAR FEP PTFE COATED. Maximum Probe Temperature Based on Coating- FEP is 200¿C and the Black PFA is 250¿C