Learn More
Savillex Corporation PFA Marking Pen, Black, 10/pk
SDP
Catalog No. NC1873960
Encompass
Black ink color. Adheres to PFA without beading or smudging. Dries in 3-6 seconds. Color-intensive, low odor. Extremely lightfast.
Catalog No. NC1873960 Supplier Savillex Corporation Supplier No. 7300400
Black ink color. Adheres to PFA without beading or smudging. Dries in 3-6 seconds. Color-intensive, low odor. Extremely lightfast.