Learn More
Perkin Elmer US LLC PFA MicroFlow High Solids Large Bore Nebulizer for Optima
Catalog No. 502104211
Encompass_Preferred
This PFA MicroFlow high solids large bore nebulizer with external 1/4-28 threaded connector is for use with Optima ICP-OES series instruments. It is ideal for strong acids, high solids, alkalis, organic solvents, soils and high salt solutions.
Catalog No. 50-210-4211 Supplier Perkin Elmer US LLC Supplier No. N0777720
This PFA MicroFlow high solids large bore nebulizer with external 1/4-28 threaded connector is for use with Optima ICP-OES series instruments. It is ideal for strong acids, high solids, alkalis, organic solvents, soils and high salt solutions.