Learn More
Glass Expansion PFA Sample Tubing 1.3mm OD x 0.18mm ID x 10m long
SDP
Catalog No. 5000555721
Encompass_Preferred
PFA Sample Tubing 1.3mm OD x 0.18mm ID x 10m long Sourced from Glass Expansion
Catalog No. 50-005-55721 Supplier Glass Expansion Supplier No. PFT018
PFA Sample Tubing 1.3mm OD x 0.18mm ID x 10m long Sourced from Glass Expansion