Learn More
Glass Expansion PFA Spray Chamber for IsoMist ROL, 50 mL cyclonic
SDP
Catalog No. 502703177
Encompass_Preferred
PFA Spray Chamber for IsoMist ROL. Replacement PFA spray chamber for ROL version IsoMist XR units.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3177 Supplier Glass Expansion Supplier No. 218092985
PFA Spray Chamber for IsoMist ROL. Replacement PFA spray chamber for ROL version IsoMist XR units.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001