Learn More
Elemental Scientific Inc PFA-ST NEBULIZER -400UL/MIN
SDP
Catalog No. NC0052422
Encompass_Preferred
MicroFlow PFA-ST self-aspirated or pumped nebulizer with external 1/4-28 threaded connector. Chemically resistant nebulizer for ICP and ICPMS. Includes two capillaries, one ES-2042 100ul/min capillary & one ES-2041 400ul/min capillary)
Catalog No. NC0052422 Supplier Elemental Scientific Inc Supplier No. ES2040
Add to Cart
Add to Cart
MicroFlow PFA-ST self-aspirated or pumped nebulizer with external 1/4-28 threaded connector. Chemically resistant nebulizer for ICP and ICPMS. Includes two capillaries, one ES-2042 100ul/min capillary & one ES-2041 400ul/min capillary)