Learn More
Perkin Elmer US LLC PFA-ST MicroFlow Concentric Nebulizer for ELAN/NexION
Catalog No. 509045565
Encompass_Preferred
MicroFlow nebulizers are constructed entirely from chemically resistant fluoropolymers, ideal for strong acids, alkalis, organics, high salt solutions and high solid concentrations. They are resistant to clogging, are reliably self-aspirated or pumped and produce a fine aerosol for high transport efficiency and high sensitivity. This nebulizer is included in all PC3 Sample Introduction Kits and in the PFA-Sapphire HF-Resistant Sample Introduction Kit (N8142002).
Catalog No. 50-904-5565 Supplier Perkin Elmer US LLC Supplier No. N8122192
MicroFlow nebulizers are constructed entirely from chemically resistant fluoropolymers, ideal for strong acids, alkalis, organics, high salt solutions and high solid concentrations. They are resistant to clogging, are reliably self-aspirated or pumped and produce a fine aerosol for high transport efficiency and high sensitivity. This nebulizer is included in all PC3 Sample Introduction Kits and in the PFA-Sapphire HF-Resistant Sample Introduction Kit (N8142002).