Learn More
Perkin Elmer US LLC PFA-ST3 MicroFlow Nebulizer for Optima 2x00/4x00/5x00/7x00/8x00 and Avio 200/500
Catalog No. NC0341730
Encompass_Preferred
The PFA MicroFlow is a high solids large bore nebulizer with external 1/4-28 threaded connector. MicroFlow nebulizers are resistant to clogging, reliably self-aspirated or pumped to produce a fine aerosol for high transport efficiency and high sensitivity. The PFA-ST3 Nebulizer is designated for superior clog resistance and higher sensitivity compared to the standard PFA nebulizer. Features and Benefits: Produces a fine aerosol for high transport efficiency and high sensitivity Low BECs even for concentrated high purity acid (e.g. 49% HF, 69% HNO3) Direct analysis of volatile and non-volatile organic solvents Ideal for VPD samples, high purity peroxide and ammonia
Catalog No. NC0341730 Supplier Perkin Elmer US LLC Supplier No. N0777676
The PFA MicroFlow is a high solids large bore nebulizer with external 1/4-28 threaded connector. MicroFlow nebulizers are resistant to clogging, reliably self-aspirated or pumped to produce a fine aerosol for high transport efficiency and high sensitivity. The PFA-ST3 Nebulizer is designated for superior clog resistance and higher sensitivity compared to the standard PFA nebulizer. Features and Benefits: Produces a fine aerosol for high transport efficiency and high sensitivity Low BECs even for concentrated high purity acid (e.g. 49% HF, 69% HNO3) Direct analysis of volatile and non-volatile organic solvents Ideal for VPD samples, high purity peroxide and ammonia