Learn More
Analytical Sales & Services PFA Teflon Tubing, 1/16" OD x .030" ID x 250'
Catalog No. 5000804737
Encompass
PFA Teflon Tubing\n\nOuter Diameter: 1/16”\nInner Diameter: 0.030”\nLength: 250’\n\nQty/Pkg: Each
Catalog No. 50-008-04737 Supplier Analytical Sales & Services Supplier No. 41030250
PFA Teflon Tubing\n\nOuter Diameter: 1/16”\nInner Diameter: 0.030”\nLength: 250’\n\nQty/Pkg: Each