Learn More
Hach Company PFA TUBING 1/4 OD X 1/8 ID 1M
Catalog No. NC2258929 Shop All Hach Company Products
Encompass
PFA tubing, 1/4 OD x 1/8 ID (6.35mm OD x 3.18mm ID)- 1 Meter
This item is not returnable. View return policy
Catalog No. NC2258929 Supplier Hach Company Supplier No. 10SCA006
PFA tubing, 1/4 OD x 1/8 ID (6.35mm OD x 3.18mm ID)- 1 Meter