Learn More
New Jersey Fluid Systems LLC PFA TUBING, 100FEET
Catalog No. NC2130620
Encompass
PFA Tubing, 1/4 in. OD x 0.062 in. Wall x 100 Feet
This item is not returnable. View return policy
Catalog No. NC2130620 Supplier New Jersey Fluid Systems LLC Supplier No. PFAT4062100
PFA Tubing, 1/4 in. OD x 0.062 in. Wall x 100 Feet