Learn More
IDEX Health & Science Upchurch PFA TUBING HI PURITY 1/16" OD
Catalog No. NC1916063
Encompass_Preferred
PFA Tubing High Purity 1/16" OD x .020" ID x 50ft
Catalog No. NC1916063 Supplier IDEX Health & Science Upchurch Supplier No. 1622L
PFA Tubing High Purity 1/16" OD x .020" ID x 50ft