Learn More
Perkin Elmer US LLC PFA Union for MEINHARD Plus Nebulizer for NexION 1000/2000
Catalog No. 502103869
Encompass_Preferred
This PFA Union is used as a fitting for MEINHARD Plus Nebulizers to adapt to the MicroFlow Concentric Nebulizer (N8152378).
Catalog No. 50-210-3869 Supplier Perkin Elmer US LLC Supplier No. N8152387
This PFA Union is used as a fitting for MEINHARD Plus Nebulizers to adapt to the MicroFlow Concentric Nebulizer (N8152378).