A 100% fluoropolymer wide-mouth wash bottle designed for ultra-pure fluid handling and storage. The PFA body and PTFE lip seals deliver low metal ion content, low extractables, and broad chemical resistance for analytical, semiconductor, pharmaceutical, and environmental laboratory use.
- 500 mL capacity (body volume to brim 515 mL).
- PFA body with PTFE lip seals.
- Wide mouth, neck I.D. 34 mm, molded-in graduations.
- Height to top of straw 290 mm; diameter 72 mm.
- Temperature resistance -196 °C to >205 °C; short-term excursions to 260 °C; microwave safe.
- Ultra-low metal ion content, no plasticizers or additives; smooth inner walls reduce contamination hang-up.
- Suitable for analytical, semiconductor, pharmaceutical, and environmental applications.
- Not intended for dispensing hazardous materials when heated or for pressurized heating during dispensing.