Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ VITLAB™ PFA Wide-Mouth Reagent Bottles

Catalog No. 14379087 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
2000 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
14-379-087 250 mL
14-379-088 500 mL
14-379-086 1000 mL
14-379-081 2000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14-379-087 Supplier BRANDTECH™ VITLAB™ Supplier No. 109497
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

VITLAB PFA wide-mouth reagent bottles are suitable for trace analysis work.

  • Leach-resistant bottles for long-term storage of standards
  • Wide mouth for easier access to contents
  • PFA screw caps help protect against outside contamination and leakage
TRUSTED_SUSTAINABILITY

Specifications

Capacity 250mL
Capacity (Metric) 250 mL
Color Translucent
Diameter (English) 2.4 in.
Material PFA
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Wide, Threaded
Height (Metric) 15 cm
Height (English) 5.9 in.
Closure Material PFA
Diameter (Metric) 6.1 cm
Closure Type Screw Cap
Autoclavable Autoclavable
Capacity (English) 8.45 oz.
Shape Sloped Shoulders
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.