Learn More
Perkin Elmer US LLC PFA-100 MicroFlow Nebulizer w/Integrated Capillary, NexION 1000/2000
Catalog No. 502104022
Encompass_Preferred
Self-aspirating PFA MicroFlow nebulizers are constructed entirely from chemically resistant fluoropolymers ideal for strong acids, alkalis, organics and high-salt solutions. They are resistant to clogging and produce a fine aerosol for high transport efficiency and high sensitivity. Benefits: Integrated capillary for self-aspiration, ideal for low flow, clean applications All PFA construction Chemically resistantideal for strong acids, alkalis, organics Low, spike-free background for important elements such as Fe and Ca Produces a fine aerosol for high transport efficiency and high sensitivity Direct analysis of volatile and non-volatile organic solvents
Catalog No. 50-210-4022 Supplier Perkin Elmer US LLC Supplier No. N8152584
Self-aspirating PFA MicroFlow nebulizers are constructed entirely from chemically resistant fluoropolymers ideal for strong acids, alkalis, organics and high-salt solutions. They are resistant to clogging and produce a fine aerosol for high transport efficiency and high sensitivity. Benefits: Integrated capillary for self-aspiration, ideal for low flow, clean applications All PFA construction Chemically resistantideal for strong acids, alkalis, organics Low, spike-free background for important elements such as Fe and Ca Produces a fine aerosol for high transport efficiency and high sensitivity Direct analysis of volatile and non-volatile organic solvents