Learn More
Perkin Elmer US LLC PFA-100 MicroFlow Nebulizer with Integrated Autosampler Probe and Ultem Support
Catalog No. 502104913
Encompass_Preferred
This nebulizer features a 100 µL/min sample uptake rate and 100 cm long Ultem Support probe for high purity semiconductor applications.
Catalog No. 50-210-4913 Supplier Perkin Elmer US LLC Supplier No. N8145143
This nebulizer features a 100 µL/min sample uptake rate and 100 cm long Ultem Support probe for high purity semiconductor applications.