Learn More
Perkin Elmer US LLC PFA-20 MicroFlow Nebulizer with Integrated Autosampler Probe - Ultem Support
Catalog No. 502111378
Encompass_Preferred
This nebulizer features a 20 µL/min sample uptake rate and 100 cm long Ultem Support probe for high purity semiconductor applications.
Catalog No. 50-211-1378 Supplier Perkin Elmer US LLC Supplier No. N8152592
This nebulizer features a 20 µL/min sample uptake rate and 100 cm long Ultem Support probe for high purity semiconductor applications.