Learn More
Perkin Elmer US LLC PFA-200 MicroFlow Nebulizer w/Integrated AS Probe - CF Support, NexION 1000/2000
Catalog No. 502111376
Encompass_Preferred
This nebulizer features a 200 µL/min sample uptake rate and 100 cm long Carbon Fiber Support probe for environmental/clinical applications.
Catalog No. 50-211-1376 Supplier Perkin Elmer US LLC Supplier No. N8152590
This nebulizer features a 200 µL/min sample uptake rate and 100 cm long Carbon Fiber Support probe for environmental/clinical applications.