Learn More
Perkin Elmer US LLC PFA-200 MicroFlow Nebulizer with Integrated Autosampler Probe and Ultem Support
Catalog No. 502104937
Encompass_Preferred
This nebulizer features a 100 µL/min sample uptake rate and 100 cm long Ultem Support probe for high purity semiconductor applications.
Catalog No. 50-210-4937 Supplier Perkin Elmer US LLC Supplier No. N8145144
Add to Cart
Add to Cart
Request a Quote
This nebulizer features a 100 µL/min sample uptake rate and 100 cm long Ultem Support probe for high purity semiconductor applications.