Learn More
Sterlitech Corporation PFA In-Line Filter Holder, 25mm
SDP
Supplier:  Sterlitech Corporation 43307010
Encompass_Preferred
Low pressure filtration (<71 psi for PP, <42 psi for PFA). Ultraclean and sterilize liquids. Aseptic sampling of liquids or gases at point-of-use. Environmental air sampling (PPO 47*).
Catalog No. NC1681610