Learn More
Perkin Elmer US LLC PFA-LC MicroFlow Nebulizer for NexION 1000/2000
Catalog No. 502103935
Encompass_Preferred
PFA LC Microflow Nebulizers, 150 µm I.D. with zero dead volume, are made from high purity, HF resistant PFA. Included are CTFE Union fittings (N8152478), a PTFE connection line (N8152479) and a capillary tube (N8152480). The sample uptake rate is controlled by the diameter of the external capillary or probe. MicroFlow nebulizers are resistant to clogging, reliably self-aspirated or pumped to produce a fine aerosol for high transport efficiency and high sensitivity.
Catalog No. 50-210-3935 Supplier Perkin Elmer US LLC Supplier No. N8152477
Add to Cart
Add to Cart
Request a Quote
PFA LC Microflow Nebulizers, 150 µm I.D. with zero dead volume, are made from high purity, HF resistant PFA. Included are CTFE Union fittings (N8152478), a PTFE connection line (N8152479) and a capillary tube (N8152480). The sample uptake rate is controlled by the diameter of the external capillary or probe. MicroFlow nebulizers are resistant to clogging, reliably self-aspirated or pumped to produce a fine aerosol for high transport efficiency and high sensitivity.