Learn More
PHENOVA PFAS CRM FOR EPA 537.1,
Catalog No. NC2231544
Encompass
Description: PFAS CRM for EPA 537.1 mix, 2 µg/mL in methanol, EaStandard Type: Calibration Standard
This item is not returnable. View return policy
Catalog No. NC2231544 Supplier PHENOVA Supplier No. AL0101839
Description: PFAS CRM for EPA 537.1 mix, 2 µg/mL in methanol, EaStandard Type: Calibration Standard