Learn More
Waters Corp PFAS Solution Installation Kit, Regional Specific, Oasis WAX 150 mg, UPLC Separation, MS Detection
Catalog No. 502391844 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit provides all necessary components for PFAS workflow needs, minimizing system-introduced PFAS contaminants using an isolator column and stainless-steel coil loop.
  • Upgrade kit type
  • One package per kit
  • UPLC separation mode
  • MS detection method
  • Room temperature storage condition
  • LC-MS system type
  • For environment applications
  • Includes separation column (1.7 μm, 2.1 × 100 mm)
  • Includes isolator column (5 μm, 2.1 × 50 mm)
  • Includes PEEK™ tubing
  • Includes PEEK™ solvent lines
  • Includes stainless-steel fittings
  • Includes solid phase extraction cartridges for PFAS analysis (6 mL/150 mg, 30/pk)
  • Includes PE/PP vials
  • Includes mobile phase bottles
Catalog No. 50-239-1844 Supplier Waters Corp Supplier No. 176004588
Add to Cart
Add to Cart
Request a Quote
This kit provides all necessary components for PFAS workflow needs, minimizing system-introduced PFAS contaminants using an isolator column and stainless-steel coil loop.
  • Upgrade kit type
  • One package per kit
  • UPLC separation mode
  • MS detection method
  • Room temperature storage condition
  • LC-MS system type
  • For environment applications
  • Includes separation column (1.7 μm, 2.1 × 100 mm)
  • Includes isolator column (5 μm, 2.1 × 50 mm)
  • Includes PEEK™ tubing
  • Includes PEEK™ solvent lines
  • Includes stainless-steel fittings
  • Includes solid phase extraction cartridges for PFAS analysis (6 mL/150 mg, 30/pk)
  • Includes PE/PP vials
  • Includes mobile phase bottles