Learn More
Waters Corp Installation Kit, Oasis WAX, 150 mg, 30 μm SPE
Catalog No. 502391136 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
PFAS Solution Installation Kits provide everything to meet your PFAS workflow needs. The kit includes necessary items to help minimize PFAS containing components within your system. Additionally, it offers a unique solution to separate any remaining system-introduced PFAS contaminants with an isolator column and stainless-steel coil loop. These kits include everything you need to get started.
  • Minimizes and separates PFAS contaminants within LC-MS systems.
  • Includes BEH C18, 1.7 μm, 2.1 x 100 mm column and BEH C18 AX, 5 μm, 2.1 x 50 mm isolator column.
  • Features WAX for PFAS analysis 6 cc Vac cartridge, 150 mg, 30 μm particle size.
  • Kit components include PEEK tubing, fittings, vials, mobile phase bottles, and essential standards.
  • Designed for UPLC separation and MS detection, compatible with EPA, ISO, and EU methods.
Catalog No. 50-239-1136 Supplier Waters Corp Supplier No. 176004548
Add to Cart
Add to Cart
Request a Quote
PFAS Solution Installation Kits provide everything to meet your PFAS workflow needs. The kit includes necessary items to help minimize PFAS containing components within your system. Additionally, it offers a unique solution to separate any remaining system-introduced PFAS contaminants with an isolator column and stainless-steel coil loop. These kits include everything you need to get started.
  • Minimizes and separates PFAS contaminants within LC-MS systems.
  • Includes BEH C18, 1.7 μm, 2.1 x 100 mm column and BEH C18 AX, 5 μm, 2.1 x 50 mm isolator column.
  • Features WAX for PFAS analysis 6 cc Vac cartridge, 150 mg, 30 μm particle size.
  • Kit components include PEEK tubing, fittings, vials, mobile phase bottles, and essential standards.
  • Designed for UPLC separation and MS detection, compatible with EPA, ISO, and EU methods.