Learn More
Perkin Elmer US LLC PFA-ST MicroFlow Nebulizer for Optima 2x00/4x00/5x00/7x00/8x00 and Avio 200/500
Catalog No. 502113243
Encompass_Preferred
MicroFlow nebulizers are resistant to clogging, reliably self-aspirated or pumped to produce a fine aerosol for high transport effciency and high sensitivity. The PFA-ST MicroFlow nebulizer is made from high purity, HF-resistant PFA. It has an exchangeable external sample uptake capillary. The sample uptake rate is controlled by the diameter of the external capillary or probe. Chemically resistantideal for strong acids, alkalis and organics, Can be pumped from < 0.1 mL/min to 3.0 mL/min, Direct analysis of volatile and non-volatile organic solvents.
Catalog No. 50-211-3243 Supplier Perkin Elmer US LLC Supplier No. N0777049
MicroFlow nebulizers are resistant to clogging, reliably self-aspirated or pumped to produce a fine aerosol for high transport effciency and high sensitivity. The PFA-ST MicroFlow nebulizer is made from high purity, HF-resistant PFA. It has an exchangeable external sample uptake capillary. The sample uptake rate is controlled by the diameter of the external capillary or probe. Chemically resistantideal for strong acids, alkalis and organics, Can be pumped from < 0.1 mL/min to 3.0 mL/min, Direct analysis of volatile and non-volatile organic solvents.