Learn More
Perkin Elmer US LLC PFA-ST Self-Aspirating Nebulizer for prepFAST Systems
Catalog No. 502105219
Encompass_Preferred
MicroFlow nebulizers are constructed entirely from chemically resistant fluoropolymers, ideal for strong acids, alkalis, organics, high salt solutions and high solid concentrations. They are resistant to clogging, are reliably self-aspirated or pumped and produce a fine aerosol for high transport efficiency and high sensitivity.
Catalog No. 50-210-5219 Supplier Perkin Elmer US LLC Supplier No. N8145368
MicroFlow nebulizers are constructed entirely from chemically resistant fluoropolymers, ideal for strong acids, alkalis, organics, high salt solutions and high solid concentrations. They are resistant to clogging, are reliably self-aspirated or pumped and produce a fine aerosol for high transport efficiency and high sensitivity.