Learn More
Perkin Elmer US LLC PFA-ST Self-Aspirating Nebulizer (PrepFAST)
Catalog No. 502111362
Encompass_Preferred
The PFA-ST MicroFlow nebulizer is made from high purity, HF resistant PFA. It has an exchangeable external sample uptake capillary. The sample uptake rate is controlled by the diameter of the external capillary or probe. Benefits: All PFA construction Exchangeable uptake capillaries allow one nebulizer to be used at different self-aspiration rates from 20 µL/min to 700 µL/min Chemically resistantideal for strong acids, alkalis and organics Can be pumped from < 0.1 mL/min to 3.0 mL/min Direct analysis of volatile and non-volatile organic solvents Longer lifetime than glass or quartz nebulizers
Catalog No. 50-211-1362 Supplier Perkin Elmer US LLC Supplier No. N8152576
The PFA-ST MicroFlow nebulizer is made from high purity, HF resistant PFA. It has an exchangeable external sample uptake capillary. The sample uptake rate is controlled by the diameter of the external capillary or probe. Benefits: All PFA construction Exchangeable uptake capillaries allow one nebulizer to be used at different self-aspiration rates from 20 µL/min to 700 µL/min Chemically resistantideal for strong acids, alkalis and organics Can be pumped from < 0.1 mL/min to 3.0 mL/min Direct analysis of volatile and non-volatile organic solvents Longer lifetime than glass or quartz nebulizers