Learn More
Perkin Elmer US LLC PFA-ST3 MicroFlow Nebulizer for NexION 1000/2000
Catalog No. 502112150
Encompass_Preferred
MicroFlow nebulizers are resistant to clogging and are reliably self-aspirated or pumped to produce a fine aerosol for high transport efficiency and high sensitivity. Benefits include: All PFA construction Longer lifetime than glass or quartz nebulizers Exchangeable uptake capillaries allow one nebulizer to be used at different self-aspiration rates from 20 µL/min to 700 µL/min Chemically resistant ideal for strong acids, alkalis and organics Can be pumped from 0.02 mL/min to 3.0 mL/min Direct analysis of volatile and non-volatile organic solvents
Catalog No. 50-211-2150 Supplier Perkin Elmer US LLC Supplier No. N8152378
MicroFlow nebulizers are resistant to clogging and are reliably self-aspirated or pumped to produce a fine aerosol for high transport efficiency and high sensitivity. Benefits include: All PFA construction Longer lifetime than glass or quartz nebulizers Exchangeable uptake capillaries allow one nebulizer to be used at different self-aspiration rates from 20 µL/min to 700 µL/min Chemically resistant ideal for strong acids, alkalis and organics Can be pumped from 0.02 mL/min to 3.0 mL/min Direct analysis of volatile and non-volatile organic solvents