Learn More
Perkin Elmer US LLC PFA-ST3 MicroFlow Nebulizer for ELAN and NexION 300/350
Catalog No. 502104869
Encompass_Preferred
This PFA-ST3 nebulizer, with external threaded connector, is a high performance, clog resistant and chemically inert nebulizer for ICP-MS applications. It has an exchangeable external sample uptake capillary where the sample uptake rate is controlled by the diameter of the external capillary or probe.
Catalog No. 50-210-4869 Supplier Perkin Elmer US LLC Supplier No. N8145101
This PFA-ST3 nebulizer, with external threaded connector, is a high performance, clog resistant and chemically inert nebulizer for ICP-MS applications. It has an exchangeable external sample uptake capillary where the sample uptake rate is controlled by the diameter of the external capillary or probe.