Learn More
TARGETMOL CHEMICALS INC PFB-FDGLU 5MG
SDP
Catalog No. 502580050
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. PFB-FDGlu is a lysosomal Glucocerebrosidase (GCase) substrate with cell-permeable specificity that is cleaved to produce fluorescein.PFB-FDGlu is often used in conjunction with a flow cytometer to measure GCase activity in living cells on an individual cell basis. purity: 99%
Catalog No. 50-258-0050 Supplier TARGETMOL CHEMICALS INC Supplier No. T395185MG
Also available in 1 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. PFB-FDGlu is a lysosomal Glucocerebrosidase (GCase) substrate with cell-permeable specificity that is cleaved to produce fluorescein.PFB-FDGlu is often used in conjunction with a flow cytometer to measure GCase activity in living cells on an individual cell basis. purity: 99%