Learn More
Accuflow Systems Saint-Gobain Pure-Fit SIB Connector, PVDF, 1/4 in
Catalog No. NC3363124
Encompass_Preferred
Smooth inner-bore connector for hygienic tubing assemblies designed to minimize dead volume and reduce contamination risk in single-use and bioprocess systems.
  • Smooth inner bore reduces dead space and turbulence.
  • PVDF construction offers chemical compatibility and temperature resistance.
  • 1/4 in (6.4 mm) inner diameter for standard tubing connections.
  • Rated to 250 psi at 72°F (22.2°C) and 100 psi at 200°F (93.3°C).
  • Hose-barb design provides secure tubing engagement.
  • Suitable for single-use and aseptic fluid transfer applications.
Catalog No. NC3363124 Supplier Accuflow Systems Supplier No. PFC250PVDF
Smooth inner-bore connector for hygienic tubing assemblies designed to minimize dead volume and reduce contamination risk in single-use and bioprocess systems.
  • Smooth inner bore reduces dead space and turbulence.
  • PVDF construction offers chemical compatibility and temperature resistance.
  • 1/4 in (6.4 mm) inner diameter for standard tubing connections.
  • Rated to 250 psi at 72°F (22.2°C) and 100 psi at 200°F (93.3°C).
  • Hose-barb design provides secure tubing engagement.
  • Suitable for single-use and aseptic fluid transfer applications.