Learn More
Accuflow Systems Saint-Gobain Pure-Fit SIB Connector, PVDF, 3/8 in, Barb x Barb
Catalog No. NC3363126
Encompass_Preferred
Saint-Gobain Pure-Fit SIB smooth inner-bore connector provides a low-dead-volume hose-barb connection for bioprocess fluid transfer, minimizing turbulence and potential contamination. The PVDF construction is autoclavable and gamma-stable, and materials meet USP Class VI requirements for bioprocess applications.
  • Smooth inner bore minimizes dead volume and turbulence.
  • Material PVDF, autoclavable and gamma stable.
  • Nominal size 3/8 in (9.50 mm), barb x barb connection.
  • Maximum pressure 250 psi at 72°F (22.2°C), 100 psi at 200°F (93.3°C).
  • USP Class VI compliant, animal-free and BPA-free materials.
Catalog No. NC3363126 Supplier Accuflow Systems Supplier No. PFC375PVDF
Saint-Gobain Pure-Fit SIB smooth inner-bore connector provides a low-dead-volume hose-barb connection for bioprocess fluid transfer, minimizing turbulence and potential contamination. The PVDF construction is autoclavable and gamma-stable, and materials meet USP Class VI requirements for bioprocess applications.
  • Smooth inner bore minimizes dead volume and turbulence.
  • Material PVDF, autoclavable and gamma stable.
  • Nominal size 3/8 in (9.50 mm), barb x barb connection.
  • Maximum pressure 250 psi at 72°F (22.2°C), 100 psi at 200°F (93.3°C).
  • USP Class VI compliant, animal-free and BPA-free materials.