Learn More
Accuflow Systems Saint-Gobain Pure-Fit SIB Cross Connector, PVDF, 1/2 in
Catalog No. NC3363122
Encompass_Preferred
The Pure-Fit SIB Cross connector is a PVDF smooth-inner-bore 4-way tubing connector engineered to provide seamless, low-dead-volume transitions for critical fluid transfer in bioprocess and laboratory systems.
  • Smooth inner-bore design minimizes dead volume and reduces fluid stagnation.
  • PVDF material offers chemical resistance and biocompatibility.
  • 1/2 in (12.7 mm) inner diameter for standard tubing compatibility.
  • Autoclavable and gamma stable for sterilization compatibility.
  • Leak-free barb interface provides secure tubing connections.
  • Typical packaging: bag of 25 (supplier packaging).
Catalog No. NC3363122 Supplier Accuflow Systems Supplier No. PFC500PVDF
The Pure-Fit SIB Cross connector is a PVDF smooth-inner-bore 4-way tubing connector engineered to provide seamless, low-dead-volume transitions for critical fluid transfer in bioprocess and laboratory systems.
  • Smooth inner-bore design minimizes dead volume and reduces fluid stagnation.
  • PVDF material offers chemical resistance and biocompatibility.
  • 1/2 in (12.7 mm) inner diameter for standard tubing compatibility.
  • Autoclavable and gamma stable for sterilization compatibility.
  • Leak-free barb interface provides secure tubing connections.
  • Typical packaging: bag of 25 (supplier packaging).