Learn More
Medchemexpress LLC HY-W078844 5mg Medchemexpress, PfDHODH-IN-2 CAS:425629-94-3 Purity:>98%
Supplier: Medchemexpress LLC HYW0788445MG
Medchemexpress, HY-W078844 5mg PfDHODH-IN-2 CAS:425629-94-3 PfDHODH-IN-2, a dihydrothiophenone derivative (Compound 11), is a potent Plasmodium falciparum dihydroorotate dehydrogenase (PfDHODH) inhibitor with an IC50 of 1.11 µM. PfDHODH-IN-2 acts as an antimalarial agent and can be used for the research of malaria. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.