Learn More
Aviva Systems Biology Corp PFDN1 Antibody - N-terminal region (ARP33025_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33025_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PFDN1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PFDN1. Alternate names: PDF, PFD1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54854