Learn More
Medchemexpress LLC HY-120085 5mg Medchemexpress, PFE-360 CAS:1527475-61-1 Purity:>98%
SDP
Catalog No. 501932547
Encompass_Preferred
Medchemexpress, HY-120085 5mg PFE-360 CAS:1527475-61-1 PFE-360 (PF-06685360) is a potent, selective, brain penetrated and orally active leucine-rich repeat kinase 2 (LRRK2) inhibitor with a mean IC 50 of 2.3 nM in vivo [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2547 Supplier Medchemexpress LLC Supplier No. HY1200855MG
Medchemexpress, HY-120085 5mg PFE-360 CAS:1527475-61-1 PFE-360 (PF-06685360) is a potent, selective, brain penetrated and orally active leucine-rich repeat kinase 2 (LRRK2) inhibitor with a mean IC 50 of 2.3 nM in vivo [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.