Learn More
Medchemexpress LLC HY-16586 10mg Medchemexpress, PFI-1 CAS:1403764-72-6 Purity:>98%
SDP
Catalog No. 502081896
Encompass_Preferred
Medchemexpress, HY-16586 10mg PFI-1 CAS:1403764-72-6 PFI-1 is a selective BET (bromodomain-containing protein) inhibitor for BRD4 with IC50 of 0.22 μM in a cell-free assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1896 Supplier Medchemexpress LLC Supplier No. HY1658610MG
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Medchemexpress, HY-16586 10mg PFI-1 CAS:1403764-72-6 PFI-1 is a selective BET (bromodomain-containing protein) inhibitor for BRD4 with IC50 of 0.22 μM in a cell-free assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.