Learn More
Selleck Chemical LLC PFI-2 HCl 10mg
SDP
Supplier:  Selleck Chemical LLC S729410MG
Encompass_Preferred
PFI-2 is a potent, selective, and cell-active lysine methyltransferase SETD7 inhibitor with Ki (app) and IC50 of 0.33 nM and 2 nM, 1000-fold selectivity over other methyltransferases and other non-epigenetic targets. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3264