Learn More
TARGETMOL CHEMICALS INC PFI-2 HYDROCHLORIDE 10MG
SDP
Catalog No. 502576748
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PFI-2 hydrochloride ((R)-PFI-2 hydrochloride) is a potent highly selective and cell-active inhibitor of the methyltransferase activity of SETD7 (IC50 2 nM) 500 fold active than (S)-PFI-2. purity: 99%
Catalog No. 50-257-6748 Supplier TARGETMOL CHEMICALS INC Supplier No. T458310MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PFI-2 hydrochloride ((R)-PFI-2 hydrochloride) is a potent highly selective and cell-active inhibitor of the methyltransferase activity of SETD7 (IC50 2 nM) 500 fold active than (S)-PFI-2. purity: 99%