Learn More
Medchemexpress LLC PFI-653 (Vanin-1-IN-1) | 2173134-00-2 | 99.9% | 25 MG
SDP
Catalog No. 5000341640
Encompass_Preferred
PFI-653 is a chemical probe and a vanin-1 inhibitor. It binds to plasma vanin-1 from mouse, rat, dog, and human. Vanin-1 is a cell surface-associated inositol (GPi)-anchored protein crucial in metabolism and inflammation.
  • Binds to plasma vanin-1 from multiple species
  • Inhibits vanin-1, a GPi-anchored protein important in metabolism and inflammation
  • Exhibits nanomolar IC50 values for vanin-1 inhibition across species
Catalog No. 50-003-41640 Supplier Medchemexpress LLC Supplier No. 25MGHY129035
PFI-653 is a chemical probe and a vanin-1 inhibitor. It binds to plasma vanin-1 from mouse, rat, dog, and human. Vanin-1 is a cell surface-associated inositol (GPi)-anchored protein crucial in metabolism and inflammation.
  • Binds to plasma vanin-1 from multiple species
  • Inhibits vanin-1, a GPi-anchored protein important in metabolism and inflammation
  • Exhibits nanomolar IC50 values for vanin-1 inhibition across species