Learn More
ATCC Pfizer Pneumococcal polysaccharide powder Type 14 (d14) 200mg
Supplier:  ATCC 81X
Encompass
Purified pneumococcal polysaccharide type 14 is manufactured by Pfizer, Inc., for small-quantity distribution by ATCC. The pneumococcal powder type 14 is a component of the current 13-valent vaccine against Streptococcus pneumoniae. The pneumococcal polysaccharide type 14 can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-2720
 Restricted Item